siRNA Details
| Gene Symbol: Ambra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134341.1 | NP_001127813.1 | 7 | 1703 | ||
| XM_008762029.1 | XP_008760251.1 | 7 | 1890 | |||
| XM_008762030.1 | XP_008760252.1 | 7 | 1607 | |||
| XM_008762031.1 | XP_008760253.1 | 6 | 1464 | |||
| XM_008762032.1 | XP_008760254.1 | 6 | 1464 | |||
| XM_008762033.1 | XP_008760255.1 | 7 | 1191 | |||
| XM_008762034.1 | XP_008760256.1 | 7 | 1191 | |||

Ambra1
Genomic Map