siRNA Details
| Gene Symbol: Kcnv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021697.1 | NP_067729.1 | 1 | 347 | ||
| GenBank mRNA | FQ211837.1 | - | 358 | |||
| X98564.1 | - | 1 | 347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnv1| Gene Symbol: Kcnv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021697.1 | NP_067729.1 | 1 | 347 | ||
| GenBank mRNA | FQ211837.1 | - | 358 | |||
| X98564.1 | - | 1 | 347 | |||