siRNA Details
| Gene Symbol: Neurog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021700.1 | NP_067732.1 | 1 | 692 | ||
| GenBank mRNA | Y10619.1 | - | 1 | 692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neurog3| Gene Symbol: Neurog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021700.1 | NP_067732.1 | 1 | 692 | ||
| GenBank mRNA | Y10619.1 | - | 1 | 692 | ||