siRNA Details
Gene Symbol: Neurog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021700.1 | NP_067732.1 | 1 | 867 | ||
GenBank mRNA | Y10619.1 | - | 1 | 867 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Neurog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021700.1 | NP_067732.1 | 1 | 867 | ||
GenBank mRNA | Y10619.1 | - | 1 | 867 |