siRNA Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031662.1 | NP_113850.1 | 16 | 1610 | ||
| XM_006246799.2 | XP_006246861.1 | 17 | 1810 | |||
| XM_006246801.2 | XP_006246863.1 | 17 | 1757 | |||
| XM_008767866.1 | XP_008766088.1 | 18 | 1899 | |||
| GenBank mRNA | AB023658.1 | - | 16 | 1610 | ||
| L42810.1 | - | 16 | 1610 | |||
| S83194.1 | - | 16 | 1610 | |||

Camkk1
Genomic Map