siRNA Details
| Gene Symbol: Csad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134454.1 | NP_001127926.1 | 3 | 297 | ||
| NM_021750.2 | NP_068518.1 | 4 | 308 | |||
| XM_006242438.2 | XP_006242500.1 | 4 | 717 | |||
| XM_006242439.1 | XP_006242501.1 | 4 | 314 | |||
| XM_006242440.2 | XP_006242502.1 | 5 | 793 | |||
| XM_006242441.2 | XP_006242503.1 | 4 | 375 | |||
| XM_006242442.2 | XP_006242504.1 | 5 | 408 | |||
| XM_008765782.1 | XP_008764004.1 | 4 | 495 | |||
| GenBank mRNA | AJ132661.1 | - | 5 | 413 | ||
| BC081804.1 | - | 4 | 308 | 7106840 | ||
| EU786150.1 | - | 3 | 280 | |||
| EU906909.1 | - | 3 | 297 | |||
| M64755.1 | - | 4 | 347 | |||
| U74492.1 | - | 2 | 158 | |||
| X94152.1 | - | 4 | 466 | |||

Csad
Genomic Map