siRNA Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021757.2 | NP_068525.1 | 9 | 795 | ||
| GenBank mRNA | AF128241.1 | - | 8 | 702 | ||
| BC087742.1 | - | 9 | 795 | 7314208 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plrg1| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021757.2 | NP_068525.1 | 9 | 795 | ||
| GenBank mRNA | AF128241.1 | - | 8 | 702 | ||
| BC087742.1 | - | 9 | 795 | 7314208 | ||