siRNA Details
| Gene Symbol: Arfip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021763.1 | NP_068531.1 | 9 | 1072 | ||
| XM_006232539.2 | XP_006232601.1 | 9 | 1333 | |||
| XM_006232540.2 | XP_006232602.1 | 10 | 1475 | |||
| XM_006232541.2 | XP_006232603.1 | 11 | 1542 | |||
| XM_006232543.1 | XP_006232605.1 | 9 | 1085 | |||
| XM_006232545.2 | XP_006232607.1 | 8 | 1236 | |||
| XM_006232546.2 | XP_006232608.1 | 9 | 1378 | |||
| XM_008761110.1 | XP_008759332.1 | 9 | 1089 | |||
| XM_008761111.1 | XP_008759333.1 | 9 | 1131 | |||
| GenBank mRNA | AY004875.1 | - | 9 | 1072 | ||
| FQ222383.1 | - | 9 | 1105 | |||

Arfip1
Genomic Map