siRNA Details
| Gene Symbol: Pfkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206847.1 | NP_996729.1 | 22 | 2373 | ||
| XM_008771885.1 | XP_008770107.1 | 22 | 2433 | |||
| GenBank mRNA | BC088847.1 | - | 22 | 2373 | 7312293 | |
| FN432820.1 | - | 22 | 2401 | |||
| L25387.1 | - | 22 | 2347 | |||

Pfkp
Genomic Map