siRNA Details
| Gene Symbol: Prmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024363.1 | NP_077339.1 | 2 | 105 | ||
| XM_006229144.2 | XP_006229206.1 | 3 | 223 | |||
| XM_006229145.2 | XP_006229207.1 | 2 | 387 | |||
| XM_006229146.2 | XP_006229208.1 | 3 | 490 | |||
| GenBank mRNA | BC078815.1 | - | 2 | 110 | 7106257 | |
| FQ213198.1 | - | 2 | 118 | |||
| FQ219843.1 | - | 2 | 116 | |||
| FQ228625.1 | - | 2 | 387 | |||
| FQ233897.1 | - | 2 | 118 | |||
| U60882.1 | - | 2 | 105 | |||

Prmt1
Genomic Map