siRNA Details
| Gene Symbol: Glp2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021848.1 | NP_068620.1 | 2 | 223 | ||
| GenBank mRNA | AF105368.1 | - | 2 | 223 | ||
| AF338223.1 | - | 2 | 327 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glp2r| Gene Symbol: Glp2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021848.1 | NP_068620.1 | 2 | 223 | ||
| GenBank mRNA | AF105368.1 | - | 2 | 223 | ||
| AF338223.1 | - | 2 | 327 | |||
Set Membership: |
|