siRNA Details
| Gene Symbol: Batf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021865.1 | NP_068637.1 | 2 | 83 | ||
| XM_008769873.1 | XP_008768095.1 | 2 | 195 | |||
| XM_008769874.1 | XP_008768096.1 | 2 | 223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Batf3| Gene Symbol: Batf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021865.1 | NP_068637.1 | 2 | 83 | ||
| XM_008769873.1 | XP_008768095.1 | 2 | 195 | |||
| XM_008769874.1 | XP_008768096.1 | 2 | 223 | |||