siRNA Details
| Gene Symbol: Hr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024364.3 | NP_077340.3 | 4 | 1334 | ||
| XM_006252283.1 | XP_006252345.1 | 1 | 561 | |||
| XM_008770833.1 | XP_008769055.1 | 1 | 561 | |||
| GenBank mRNA | U71293.1 | - | 1 | 950 | ||

Hr
Genomic Map