siRNA Details
| Gene Symbol: Npvf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023952.1 | NP_076442.1 | 2 | 339 | ||
| GenBank mRNA | AB040288.1 | - | 2 | 339 | ||
| AF330059.2 | - | 2 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npvf| Gene Symbol: Npvf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023952.1 | NP_076442.1 | 2 | 339 | ||
| GenBank mRNA | AB040288.1 | - | 2 | 339 | ||
| AF330059.2 | - | 2 | 339 | |||