siRNA Details
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764322.1 | XP_008762544.1 | 17 | 2925 | ||
GenBank mRNA | X59290.1 | - | 7 | 981 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764322.1 | XP_008762544.1 | 17 | 2925 | ||
GenBank mRNA | X59290.1 | - | 7 | 981 |