siRNA Details
| Gene Symbol: Ppp2r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022209.3 | NP_071545.2 | 1 | 214 | ||
| GenBank mRNA | AY251277.1 | - | 1 | 166 | ||
| FM080850.1 | - | 1 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r2b| Gene Symbol: Ppp2r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022209.3 | NP_071545.2 | 1 | 214 | ||
| GenBank mRNA | AY251277.1 | - | 1 | 166 | ||
| FM080850.1 | - | 1 | 214 | |||