siRNA Details
Gene Symbol: Cxcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022214.1 | NP_071550.1 | 3 | 295 | ||
GenBank mRNA | U90448.1 | - | 3 | 295 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cxcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022214.1 | NP_071550.1 | 3 | 295 | ||
GenBank mRNA | U90448.1 | - | 3 | 295 |