siRNA Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022220.2 | NP_071556.2 | 3 | 250 | ||
| XM_006252146.2 | XP_006252208.1 | 3 | 391 | |||
| GenBank mRNA | BC089803.1 | - | 3 | 250 | 7324344 | |
| FQ219140.1 | - | 3 | 275 | |||
| FQ219632.1 | - | 3 | 358 | |||

Gulo
Genomic Map