siRNA Details
| Gene Symbol: Entpd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033565.1 | NP_001028737.1 | 4, 5 | 561 | ||
| XM_006233639.2 | XP_006233701.1 | 5, 6 | 833 | |||
| XM_006233640.2 | XP_006233702.1 | 6, 7 | 927 | |||
| XM_006233642.2 | XP_006233704.1 | 5, 6 | 818 | |||
| XM_006233643.2 | XP_006233705.1 | 5, 6 | 833 | |||
| GenBank mRNA | AY536920.1 | - | 4, 5 | 561 | ||

Entpd8
Genomic Map