siRNA Details
| Gene Symbol: Entpd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033565.1 | NP_001028737.1 | 6 | 1002 | ||
| XM_006233639.2 | XP_006233701.1 | 7 | 1274 | |||
| XM_006233640.2 | XP_006233702.1 | 8 | 1368 | |||
| XM_006233642.2 | XP_006233704.1 | 7 | 1259 | |||
| XM_006233643.2 | XP_006233705.1 | 7 | 1274 | |||
| GenBank mRNA | AY536920.1 | - | 6 | 1002 | ||

Entpd8
Genomic Map