siRNA Details
| Gene Symbol: Jam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034004.1 | NP_001029176.1 | 6 | 891 | ||
| GenBank mRNA | BC105305.1 | - | 6 | 891 | 7192610 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jam2| Gene Symbol: Jam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034004.1 | NP_001029176.1 | 6 | 891 | ||
| GenBank mRNA | BC105305.1 | - | 6 | 891 | 7192610 | |