siRNA Details
| Gene Symbol: Ncoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034007.1 | NP_001029179.1 | 8 | 1755 | ||
| NM_001034008.1 | NP_001029180.1 | 8 | 1801 | |||
| XM_006252660.2 | XP_006252722.1 | 8 | 2102 | |||
| GenBank mRNA | BC160817.1 | - | 8 | 1780 | 7454028 | |

Ncoa4
Genomic Map