siRNA Details
| Gene Symbol: Dnajc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034032.1 | NP_001029204.1 | 3 | 282 | ||
| GenBank mRNA | AB062135.1 | - | 3 | 284 | ||
| BC100072.1 | - | 3 | 282 | 7325259 | ||
| FQ214972.1 | - | 3 | 295 | |||
| FQ219445.1 | - | 3 | 295 | |||

Dnajc12
Genomic Map