siRNA Details
Gene Symbol: Med24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034079.1 | NP_001029251.1 | 26 | 2955 | ||
XM_006247415.2 | XP_006247477.1 | 27 | 3052 | |||
XM_006247416.2 | XP_006247478.1 | 27 | 3098 | |||
GenBank mRNA | BC097461.1 | - | 26 | 2955 | 7457533 | |
FQ210692.1 | - | 26 | 2855 |
