siRNA Details
| Gene Symbol: Med24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034079.1 | NP_001029251.1 | 9 | 933 | ||
| XM_006247415.2 | XP_006247477.1 | 10 | 1030 | |||
| XM_006247416.2 | XP_006247478.1 | 10 | 1076 | |||
| GenBank mRNA | BC097461.1 | - | 9 | 933 | 7457533 | |
| FQ210692.1 | - | 9 | 956 | |||

Med24
Genomic Map