siRNA Details
| Gene Symbol: Aarsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034109.1 | NP_001029281.1 | 7 | 727 | ||
| GenBank mRNA | BC083790.1 | - | 7 | 727 | 7106366 | |
| FQ213285.1 | - | 7 | 728 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aarsd1| Gene Symbol: Aarsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034109.1 | NP_001029281.1 | 7 | 727 | ||
| GenBank mRNA | BC083790.1 | - | 7 | 727 | 7106366 | |
| FQ213285.1 | - | 7 | 728 | |||