siRNA Details
| Gene Symbol: Sohlh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034961.1 | NP_001030133.1 | 2 | 226 | ||
| GenBank mRNA | BC104697.1 | - | 2 | 226 | 7113652 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sohlh2| Gene Symbol: Sohlh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034961.1 | NP_001030133.1 | 2 | 226 | ||
| GenBank mRNA | BC104697.1 | - | 2 | 226 | 7113652 | |