siRNA Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034997.1 | NP_001030169.1 | 8, 9 | 830 | ||
| GenBank mRNA | BC104694.1 | - | 8, 9 | 830 | 7110947 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrbp1| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034997.1 | NP_001030169.1 | 8, 9 | 830 | ||
| GenBank mRNA | BC104694.1 | - | 8, 9 | 830 | 7110947 | |