siRNA Details
| Gene Symbol: Ctps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034998.2 | NP_001030170.1 | 16 | 1561 | ||
| XM_006256895.2 | XP_006256957.1 | 16 | 1563 | |||
| XM_006256896.2 | XP_006256958.1 | 16 | 1851 | |||
| XM_006256897.2 | XP_006256959.1 | 16 | 1684 | |||
| XM_006256898.2 | XP_006256960.1 | 16 | 1645 | |||
| XM_006256899.2 | XP_006256961.1 | 16 | 2016 | |||
| XM_006256900.2 | XP_006256962.1 | 16 | 1659 | |||
| GenBank mRNA | BC085358.1 | - | 9 | 663 | ||
| BC085949.1 | - | 16 | 1561 | 7132135 | ||

Ctps2
Genomic Map