siRNA Details
Gene Symbol: Per2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031678.1 | NP_113866.1 | 3, 4 | 399 | ||
XM_006245476.2 | XP_006245538.1 | 3, 4 | 634 | |||
XM_006245477.2 | XP_006245539.1 | 3, 4 | 550 | |||
GenBank mRNA | AB016532.1 | - | 3, 4 | 399 |
