siRNA Details
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031684.2 | NP_113872.1 | 13, 14 | 1374 | ||
XM_006244560.1 | XP_006244622.1 | 13, 14 | 1405 | |||
XM_006244561.2 | XP_006244623.1 | 14, 15 | 1638 | |||
XM_006244562.2 | XP_006244624.1 | 13, 14 | 1577 | |||
XM_006244563.2 | XP_006244625.1 | 13, 14 | 1400 | |||
XM_008766879.1 | XP_008765101.1 | 14, 15 | 1463 | |||
GenBank mRNA | AF015304.1 | - | 10, 11 | 1060 | ||
BC078789.1 | - | 13, 14 | 1374 | 7132212 |
