siRNA Details
| Gene Symbol: Slc29a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031684.2 | NP_113872.1 | 7 | 756 | ||
| XM_006244560.1 | XP_006244622.1 | 7 | 787 | |||
| XM_006244561.2 | XP_006244623.1 | 8 | 1020 | |||
| XM_006244562.2 | XP_006244624.1 | 7 | 959 | |||
| XM_006244563.2 | XP_006244625.1 | 7 | 782 | |||
| XM_008766879.1 | XP_008765101.1 | 8 | 845 | |||
| GenBank mRNA | AF015304.1 | - | 4 | 442 | ||
| BC078789.1 | - | 7 | 756 | 7132212 | ||

Slc29a1
Genomic Map