siRNA Details
| Gene Symbol: Dusp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022248.1 | NP_071584.1 | 1, 2 | 349 | ||
| GenBank mRNA | AF217233.1 | - | 1, 2 | 349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp12| Gene Symbol: Dusp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022248.1 | NP_071584.1 | 1, 2 | 349 | ||
| GenBank mRNA | AF217233.1 | - | 1, 2 | 349 | ||