siRNA Details
| Gene Symbol: Enpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022251.2 | NP_071587.2 | 1 | 624 | ||
| GenBank mRNA | AF214568.1 | - | 1 | 441 | ||
| BC066663.1 | - | 1 | 446 | 5623429 | ||
| CK481363.1 | - | 464 | ||||
| Gene Symbol: LOC100910501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761484.1 | XP_008759706.1 | 1 | 472 | ||

Enpep
Genomic Map