siRNA Details
Gene Symbol: Bspry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022261.2 | NP_071597.2 | 5 | 782 | ||
GenBank mRNA | AF245225.1 | - | 3 | 309 | ||
BC062051.1 | - | 5 | 730 | |||
BC095901.1 | - | 5 | 433 | |||
BC111380.1 | - | 4 | 336 | |||
BC166587.1 | - | 5 | 782 | 7937509 |
