siRNA Details
| Gene Symbol: Tsnax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022262.3 | NP_071598.1 | 6 | 757 | ||
| GenBank mRNA | AF262357.1 | - | 6 | 828 | ||
| BC081715.1 | - | 6 | 757 | 7130295 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsnax| Gene Symbol: Tsnax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022262.3 | NP_071598.1 | 6 | 757 | ||
| GenBank mRNA | AF262357.1 | - | 6 | 828 | ||
| BC081715.1 | - | 6 | 757 | 7130295 | ||