siRNA Details
| Gene Symbol: Cd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022269.2 | NP_071605.2 | 7 | 1038 | ||
| GenBank mRNA | AB026901.1 | - | 7 | 982 | ||
| AB026903.1 | - | 7 | 982 | |||
| AB026904.1 | - | 6 | 831 | |||
| AF039583.1 | - | 7 | 979 | |||
| BC061869.1 | - | 7 | 1002 | 5598916 | ||

Cd55
Genomic Map