siRNA Details
| Gene Symbol: Hapln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022285.1 | NP_071621.1 | 7 | 1035 | ||
| XM_006232751.2 | XP_006232813.1 | 7 | 1117 | |||
| XM_008761208.1 | XP_008759430.1 | 8 | 1158 | |||
| XM_008761209.1 | XP_008759431.1 | 7 | 1343 | |||
| GenBank mRNA | AB049056.1 | - | 7 | 1035 | ||

Hapln2
Genomic Map