siRNA Details
| Gene Symbol: Hapln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022285.1 | NP_071621.1 | 3 | 250 | ||
| XM_006232751.2 | XP_006232813.1 | 3 | 332 | |||
| XM_008761208.1 | XP_008759430.1 | 4 | 373 | |||
| XM_008761209.1 | XP_008759431.1 | 3 | 558 | |||
| GenBank mRNA | AB049056.1 | - | 3 | 250 | ||

Hapln2
Genomic Map