siRNA Details
| Gene Symbol: Atp5h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019383.2 | NP_062256.1 | 2 | 130 | ||
| XM_008768390.1 | XP_008766612.1 | 2 | 167 | |||
| GenBank mRNA | BC059139.1 | - | 2 | 134 | 6921557 | |
| BC078846.1 | - | 1 | 82 | 7133871 | ||
| D10021.1 | - | 1 | 102 | |||
| D13120.1 | - | 2 | 123 | |||
| FQ211219.1 | - | 2 | 141 | |||
| FQ216885.1 | - | 2 | 130 | |||
| FQ217843.1 | - | 2 | 141 | |||
| FQ222479.1 | - | 2 | 123 | |||

Atp5h
Genomic Map