siRNA Details
Gene Symbol: Lrrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037336.1 | NP_001032413.1 | 1 | 978 | ||
GenBank mRNA | DQ119102.1 | - | 1 | 978 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037336.1 | NP_001032413.1 | 1 | 978 | ||
GenBank mRNA | DQ119102.1 | - | 1 | 978 |