siRNA Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037664.1 | NP_001032753.1 | 5, 6 | 458 | ||
| GenBank mRNA | AY623025.1 | - | 1931 | |||
| AY623026.1 | - | 1032 | ||||
| BC088267.1 | - | 5, 6 | 458 | 7379090 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtnbp1| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037664.1 | NP_001032753.1 | 5, 6 | 458 | ||
| GenBank mRNA | AY623025.1 | - | 1931 | |||
| AY623026.1 | - | 1032 | ||||
| BC088267.1 | - | 5, 6 | 458 | 7379090 | ||