siRNA Details
Gene Symbol: Uba52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031687.3 | NP_113875.1 | 2, 3 | 142 | ||
XM_006252934.2 | XP_006252996.1 | 2, 3 | 229 | |||
XM_006252935.1 | XP_006252997.1 | 2, 3 | 347 | |||
XM_008771145.1 | XP_008769367.1 | 2, 3 | 225 | |||
XM_008771146.1 | XP_008769368.1 | 2, 3 | 169 | |||
GenBank mRNA | BC061544.1 | - | 2, 3 | 138 | 6922034 | |
BP480615.1 | - | 2, 3 | 338 | |||
BP492585.1 | - | 1, 2 | 28 | |||
FQ209944.1 | - | 2, 3 | 134 | |||
FQ210541.1 | - | 2, 3 | 131 | |||
FQ210904.1 | - | 2, 3 | 132 | |||
FQ212001.1 | - | 2, 3 | 136 | |||
FQ212205.1 | - | 2, 3 | 134 | |||
FQ212291.1 | - | 2, 3 | 132 | |||
FQ212810.1 | - | 2, 3 | 135 | |||
FQ217440.1 | - | 2, 3 | 127 | |||
FQ218121.1 | - | 2, 3 | 128 | |||
FQ219376.1 | - | 2, 3 | 133 | |||
FQ219803.1 | - | 2, 3 | 134 | |||
FQ221045.1 | - | 2, 3 | 131 | |||
FQ221118.1 | - | 2, 3 | 132 | |||
FQ221238.1 | - | 2, 3 | 352 | |||
FQ221621.1 | - | 2, 3 | 128 | |||
FQ221631.1 | - | 2, 3 | 134 | |||
FQ221755.1 | - | 2, 3 | 132 | |||
FQ221772.1 | - | 2, 3 | 132 | |||
FQ221777.1 | - | 2, 3 | 132 | |||
FQ222045.1 | - | 2, 3 | 132 | |||
FQ222098.1 | - | 2, 3 | 128 | |||
FQ222371.1 | - | 2, 3 | 131 | |||
FQ222440.1 | - | 2, 3 | 143 | |||
FQ222483.1 | - | 2, 3 | 131 | |||
FQ222507.1 | - | 2, 3 | 126 | |||
FQ222700.1 | - | 2, 3 | 131 | |||
FQ223258.1 | - | 2, 3 | 134 | |||
FQ223423.1 | - | 2, 3 | 128 | |||
FQ223456.1 | - | 2, 3 | 129 | |||
FQ224647.1 | - | 1, 2 | 122 | |||
FQ224702.1 | - | 2, 3 | 132 | |||
FQ228362.1 | - | 2, 3 | 128 | |||
FQ228566.1 | - | 2, 3 | 132 | |||
X82636.1 | - | 1, 2 | 104 |
