siRNA Details
| Gene Symbol: Ascl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022384.1 | NP_071779.1 | 1 | 1354 | ||
| GenBank mRNA | X53725.1 | - | 1 | 1354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ascl1| Gene Symbol: Ascl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022384.1 | NP_071779.1 | 1 | 1354 | ||
| GenBank mRNA | X53725.1 | - | 1 | 1354 | ||