siRNA Details
| Gene Symbol: Clec10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022393.2 | NP_071788.2 | 4 | 511 | ||
| XM_006246803.2 | XP_006246865.1 | 4 | 549 | |||
| XM_008767867.1 | XP_008766089.1 | 4 | 549 | |||
| XM_008767868.1 | XP_008766090.1 | 4 | 393 | |||
| XM_008767869.1 | XP_008766091.1 | 4 | 390 | |||
| GenBank mRNA | BC091151.1 | - | 4 | 511 | 7375234 | |
| FQ213042.1 | - | 4 | 383 | |||
| FQ221204.1 | - | 4 | 400 | |||
| FQ228437.1 | - | 4 | 369 | |||
| J05495.1 | - | 4 | 371 | |||

Clec10a
Genomic Map