siRNA Details
| Gene Symbol: Bcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022400.1 | NP_071795.1 | 4, 5 | 430 | ||
| XM_006229149.1 | XP_006229211.1 | 4, 5 | 607 | |||
| GenBank mRNA | U68417.1 | - | 4, 5 | 430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcat2| Gene Symbol: Bcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022400.1 | NP_071795.1 | 4, 5 | 430 | ||
| XM_006229149.1 | XP_006229211.1 | 4, 5 | 607 | |||
| GenBank mRNA | U68417.1 | - | 4, 5 | 430 | ||