siRNA Details
| Gene Symbol: Mthfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022508.1 | NP_071953.1 | 3 | 447 | ||
| XM_006240249.2 | XP_006240311.1 | 3 | 586 | |||
| GenBank mRNA | BC089800.1 | - | 3 | 161 | 7323085 | |
| J05519.1 | - | 3 | 447 | |||
| Gene Symbol: LOC680432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762232.1 | XP_008760454.1 | 1 | 168 | ||

Mthfd1
Genomic Map