siRNA Details
| Gene Symbol: Mthfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022508.1 | NP_071953.1 | 5 | 614 | ||
| GenBank mRNA | J05519.1 | - | 5 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mthfd1| Gene Symbol: Mthfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022508.1 | NP_071953.1 | 5 | 614 | ||
| GenBank mRNA | J05519.1 | - | 5 | 614 | ||