siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022513.2 | NP_071958.1 | 2 | 307 | ||
| XM_008770056.1 | XP_008768278.1 | 2 | 181 | |||
| GenBank mRNA | D89375.1 | - | 2 | 258 | ||
| FQ209476.1 | - | 2 | 276 | |||
| FQ209486.1 | - | 2 | 341 | |||
| FQ209579.1 | - | 2 | 362 | |||
| FQ210801.1 | - | 2 | 349 | |||
| FQ212608.1 | - | 2 | 318 | |||
| FQ218295.1 | - | 2 | 320 | |||
| U38419.1 | - | 2 | 310 | |||

Sult1b1
Genomic Map