siRNA Details
| Gene Symbol: Dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022526.2 | NP_071971.1 | 4 | 483 | ||
| GenBank mRNA | BC060569.1 | - | 4 | 301 | 6890457 | |
| FQ218203.1 | - | 4 | 368 | |||
| FQ228698.1 | - | 4 | 321 | |||
| FQ228865.1 | - | 4 | 327 | |||
| U05334.1 | - | 4 | 484 | |||

Dap
Genomic Map